ContactUsLinkComponent
RECOMBINANT/NATIVE PROTEIN, HA1 (H2N2) (A/CANADA/720/2005) RECOMBINANT PROTEIN, HA1 (H2N2) (A/CANADA/720/2005) (AAY28987, 13 A.A. - 340 A.A.) PARTIAL RECOMBINANT PROTEIN WITH HIS TAG EXPRESSED IN 293 CELLS. 1 * 50 µG
Catalog # ABNOP5044
Supplier:  Abnova
RECOMBINANT/NATIVE PROTEIN, HA1 (H2N2) (A/CANADA/720/2005) RECOMBINANT PROTEIN, HA1 (H2N2) (A/CANADA/720/2005) (AAY28987, 13 A.A. - 340 A.A.) PARTIAL RECOMBINANT PROTEIN WITH HIS TAG EXPRESSED IN 293 CELLS. 1 * 50 µG
Catalog # ABNOP5044
Supplier:  Abnova