ContactUsLinkComponent
RECOMBINANT/NATIVE PROTEIN, ADH1A (HUMAN) RECOMBINANT PROTEIN, HUMAN ADH1A (NP_000658, 1 A.A. - 375 A.A.) FULL-LENGTH RECOMBINANT PR OTEIN WITH HIS TAG EXPRESSED IN ESCHERICHIA COLI. 1 * 100 µG
Catalog # ABNOP5012
Supplier:  Abnova
RECOMBINANT/NATIVE PROTEIN, ADH1A (HUMAN) RECOMBINANT PROTEIN, HUMAN ADH1A (NP_000658, 1 A.A. - 375 A.A.) FULL-LENGTH RECOMBINANT PR OTEIN WITH HIS TAG EXPRESSED IN ESCHERICHIA COLI. 1 * 100 µG
Catalog # ABNOP5012
Supplier:  Abnova