ContactUsLinkComponent
KIT, HYDROGEN PEROXIDE COLORIMETRIC DETECTION KIT, HYDROGEN PEROXIDE COLORIMETRIC DETECTION KIT IS A HIGHER THROUGHPUT ASSAY FOR THE QUANTITATIVE MEASUREMENT OF HYDROGEN PEROXIDE IN CULTURE SUPERNATANTS, SERUM, PLASMA, AND URINE FROM ANY SPECIES. 1 * 1 KIT
Catalog # ABNOKA1017
Supplier:  Abnova
KIT, HYDROGEN PEROXIDE COLORIMETRIC DETECTION KIT, HYDROGEN PEROXIDE COLORIMETRIC DETECTION KIT IS A HIGHER THROUGHPUT ASSAY FOR THE QUANTITATIVE MEASUREMENT OF HYDROGEN PEROXIDE IN CULTURE SUPERNATANTS, SERUM, PLASMA, AND URINE FROM ANY SPECIES. 1 * 1 KIT
Catalog # ABNOKA1017
Supplier:  Abnova
Frequently Bought Together