KIT, SCGB1A1 (HUMAN) ELISA KIT, SCGB1A1 (HUMAN) ELISA KIT IS A BIOTIN LABELLED ANTIBODY BASED SANDWICH ENZYME IMMUNOASSAY FOR THE QU ANTITATIVE MEASUREMENT OF HUMAN CLARA CELL PROTEIN IN SERUM, PLASMA, TISSUE CULTURE MEDIUM AND BRONCHOALVEOLAR LAVAGE 1 * 1 KIT
Catalog # ABNOKA0020
Supplier: Abnova
Frequently Bought Together