ContactUsLinkComponent
PRODUCT FOR DNA RELATE APPLICATION, DNA MARKER HIGH, DNA MARKER HIGH IS USEFUL FOR DETERMINATION OF DNA FRAGMENTS 300 BP TO 10,000 B P IN SIZE. THE DNA SIZE MARKER CONSISTS OF 13 DOUBLE-STRANDED, BLUNT-END DNA FRAGMENTS. THE DNA BANDS CLOSE TO EACH O 1 * 1 mL
Catalog # ABNOD0002
Supplier:  Abnova
PRODUCT FOR DNA RELATE APPLICATION, DNA MARKER HIGH, DNA MARKER HIGH IS USEFUL FOR DETERMINATION OF DNA FRAGMENTS 300 BP TO 10,000 B P IN SIZE. THE DNA SIZE MARKER CONSISTS OF 13 DOUBLE-STRANDED, BLUNT-END DNA FRAGMENTS. THE DNA BANDS CLOSE TO EACH O 1 * 1 mL
Catalog # ABNOD0002
Supplier:  Abnova
Frequently Bought Together