ContactUsLinkComponent
PRODUCT FOR DNA RELATE APPLICATION, DNA MARKER LOW, DNA MARKER LOW IS USEFUL FOR DETERMINATION OF DNA FRAGMENTS 50 BP TO 1,000 BP IN SIZE. THE DNA SIZE MARKER CONSISTS OF 10 DOUBLE-STRANDED, BLUNT-END DNA FRAGMENTS. THE DNA BANDS CLOSE TO EACH OTHER 1 * 1 mL
Catalog # ABNOD0001
Supplier:  Abnova
PRODUCT FOR DNA RELATE APPLICATION, DNA MARKER LOW, DNA MARKER LOW IS USEFUL FOR DETERMINATION OF DNA FRAGMENTS 50 BP TO 1,000 BP IN SIZE. THE DNA SIZE MARKER CONSISTS OF 10 DOUBLE-STRANDED, BLUNT-END DNA FRAGMENTS. THE DNA BANDS CLOSE TO EACH OTHER 1 * 1 mL
Catalog # ABNOD0001
Supplier:  Abnova
Frequently Bought Together