30 COMPOSES A 100 MG/KG DANS HNO3 5% Al As Ba Bi Ca Cd Co Cr Cu Fe Ga K Li Mg Mn Mo Na Nb Ni P Pb Sb Se Si Sn Sr Ti V W Zn 1 * 100 mL
Catalog # CMPLF386.K1.5N.L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together