Transfer electrophoresis unit TE22 Mini transfer tank Mighty small 1 * 1 items
732-2123
Transfer electrophoresis unit TE22 Mini transfer tank Mighty small 1 * 1 items
732-2123
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Pipette tips, Finntip™
: Thermo Fisher Scientific
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Assistent Brushes with Cotton Tip Bristles
: GLASWARENFABRIK KARL HECHT
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Ethanol absolute ≥99.5% USP, Ph. Eur.
: VWR Chemicals
Ethanol 96% vol Ph. Eur.
: VWR Chemicals
VWR® Standard Line Transfer Pipettes
: VWR Collection
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
3-in-1 pH/ATC probes, Orion™ Triode™
: Thermo Orion