Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
S-Ethylisothiouronium bromide

S-Ethylisothiouronium bromide

Glass bottle for solids, 1 g
Katalogové číslo CAYM81275-1
Dodavatel:   Cayman Chemical
S-Ethylisothiouronium bromide
Katalogové číslo CAYM81275-1
Dodavatel:  Cayman Chemical
Číslo CAS:  1071-37-0
S-Ethylisothiouronium bromide

Glass bottle for solids, 1 g

Přejít na:
Číslo CAS:  1071-37-0
Price
8 460,00 Kč
Přihlásit se pro dostupnost
1 * 1 g
Product Details & Documents
Synonyma:  Ethyl carbamimidothioate hydrobromide, S-Ethylisothiourea Hydrobromide, S-EIT hydrobromide, 2-Ethyl-2-thiopseudourea hydrobromide
  • Číslo CAS:1071-37-0
  • Vzorec:C₃H₉BrN₂S
  • MDL číslo:MFCD00012585
  • Molecular Weight:185.1 g/mol
Specifikace

Specifikace

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifikace

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.