Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Buffer, HEPES solution (1 mol/L)

Buffer, HEPES solution (1 mol/L)

HEPES buffer solution (1 mol/l), Glass bottle, 500 mL
Catalog # CAYM600212-500
Supplier:   Cayman Chemical
Buffer, HEPES solution (1 mol/L)
Catalog # CAYM600212-500
Supplier:  Cayman Chemical
CAS Number:  
Buffer, HEPES solution (1 mol/L)

HEPES buffer solution (1 mol/l), Glass bottle, 500 mL

CAS Number:  
Price
3 200,00 Kč
Log in for availability
1 * 500 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    HEPES buffer solution (1 mol/l)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.