Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
 

Buffer, HEPES solution (1 mol/L)

HEPES buffer solution (1 mol/l), Glass bottle, 250 ml
Catalog # CAYM600212-250
Supplier:   Cayman Chemical
Buffer, HEPES solution (1 mol/L)
Catalog # CAYM600212-250
Supplier:  Cayman Chemical
CAS Number:  
Buffer, HEPES solution (1 mol/L)

HEPES buffer solution (1 mol/l), Glass bottle, 250 ml

CAS Number:  
Price
2 740,00 Kč
Log in for availability
1 * 250 mL
Product Details & Documents

 

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 ml
  • Description:
    HEPES buffer solution (1 mol/l)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.