Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Buffer, HEPES solution (1 mol/L)

Buffer, HEPES solution (1 mol/L)

HEPES buffer solution (1 mol/l), Glass bottle, 100 ml
Catalog # CAYM600212-100
Supplier:   Cayman Chemical
Buffer, HEPES solution (1 mol/L)
Catalog # CAYM600212-100
Supplier:  Cayman Chemical
CAS Number:  
Buffer, HEPES solution (1 mol/L)

HEPES buffer solution (1 mol/l), Glass bottle, 100 ml

CAS Number:  
Price
1 480,00 Kč
Log in for availability
1 * 100 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 ml
  • Description:
    HEPES buffer solution (1 mol/l)

Specifications