STARTERKIT W. USB ADAPTER f. Datalogger DELTA T USB Starter kit To read out ThermoScan Datalogger on your PC the USB-Kit is required. It is simply plugged into one of your USB ports and the ThermoScan logger needs to be placed into the BlueDot-Recept 1 * 1 SET
Catalog # DLTTD100010DE
Supplier: DELTA T
Frequently Bought Together