ContactUsLinkComponent
29 COMPOSES DANS ACETONE (Ampoules de 1 ml ) 1-(3,4-Dichlorophenyl)-3-methylurea [CAS:3567-62-2] 10mg/l Azinphos-ethyl [CAS:2642-71-9] 25mg/l Azinphos-methyl [CAS:86-50-0] 25mg/l Bifenox [CAS:42576-02-3] 25mg/l Chlorfenvinphos [CAS:470-90-6] 1 * 120 Ampoul
Catalog # CMPLD80C.25.A.1
29 COMPOSES DANS ACETONE (Ampoules de 1 ml ) 1-(3,4-Dichlorophenyl)-3-methylurea [CAS:3567-62-2] 10mg/l Azinphos-ethyl [CAS:2642-71-9] 25mg/l Azinphos-methyl [CAS:86-50-0] 25mg/l Bifenox [CAS:42576-02-3] 25mg/l Chlorfenvinphos [CAS:470-90-6] 1 * 120 Ampoul
Catalog # CMPLD80C.25.A.1
Frequently Bought Together