ContactUsLinkComponent
CRYOBANK system for freezing bacterial cultures for later retrieval. CRYOBANK is based on a cryovial system that contains 25 color-coded chemically treated ceramic beads suspended in a special cryogenic preserving solution. The special cryogenic me 1 * 64 items
Catalog # COPACRYO/M
Supplier:  Copan
CRYOBANK system for freezing bacterial cultures for later retrieval. CRYOBANK is based on a cryovial system that contains 25 color-coded chemically treated ceramic beads suspended in a special cryogenic preserving solution. The special cryogenic me 1 * 64 items
Catalog # COPACRYO/M
Supplier:  Copan
Frequently Bought Together