ContactUsLinkComponent
868mhz wireless radio module system, differential pressure sensor, range -500 to +500 pa. (typical application: clean room) package consists of: module, 3.6v lithium battery, mounting holder w/ velcro, magnet & hardware, cd rom with software drivers 1 * 1 items
Catalog # THBRSV208-201-LSB
868mhz wireless radio module system, differential pressure sensor, range -500 to +500 pa. (typical application: clean room) package consists of: module, 3.6v lithium battery, mounting holder w/ velcro, magnet & hardware, cd rom with software drivers 1 * 1 items
Catalog # THBRSV208-201-LSB
Frequently Bought Together