ContactUsLinkComponent
868mhz wireless radio module system, external temperature probe, 25 cm in length, range -40#c to +120#c, (typical application: incu bator or water bath). package consists of: module, external sensor, 3.6v lithium battery, mounting holder w/ velcro, m 1 * 1 items
Catalog # THBRSV205-201-LSB
868mhz wireless radio module system, external temperature probe, 25 cm in length, range -40#c to +120#c, (typical application: incu bator or water bath). package consists of: module, external sensor, 3.6v lithium battery, mounting holder w/ velcro, m 1 * 1 items
Catalog # THBRSV205-201-LSB
Frequently Bought Together