13 COMPOSES DANS HNO3 2% Ca 150mg/l Mg 50mg/l K 10mg/l Na 100mg/l P 0.5mg/l Mn 0.2mg/l Fe 0.2mg/l Cr 0.2mg/l Al 0.2mg/l Cu 0.2mg/l Ni 0.2mg/l B 0.5mg/l Zn 0.5mg/l 1 * 1 L
Catalog # CMPL12E6.D2.2N.1L
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together