CONTAINER 200ML W/ CAP PP IRRAD 1 * 200 items
Catalog # 300-0133
Supplier: Anicrin
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
Accessories for Magnetic Hotplate Stirrers
Supplier: IKA
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
SOLUTION CARREZ 2 (ZINC SALT) 1 * 1 L
Supplier: PROLABO CMP
Test kits, nitrate, Spectroquant®, Supelco®
Supplier: Merck
L(+)-Potassium hydrogen tartrate 99.5-100.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR®, Sampling Spoons
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
McFARLAND 0.5 BARIUM SULPHATE STANDARD 1 * 1 items
Supplier: LIOFILCHEM