Applicators
Sterile swabs, Aluminium/cotton, 150 mm, 0,9 – 1,0 mm
Catalog # HERE1032648
Supplier: HERENZ HEINZ
Jump to:
Product Details & Documents
Aluminium applicators with a single cotton bud.
: In a single peel-pack.
Specifications
- Ø:0,9 – 1,0 mm
- Description:Sterile swabs
- Length:150 mm
- Shaft/head:Aluminium/cotton
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



