ContactUsLinkComponent
2 COMPOSES A 1 G/L CHAQUE DANS H2O ( Ampoules de 10 ml ) of Formaldehyde [CAS:50-00-0] Acetaldehyde [CAS:75-07-0] 1 * 1 Ampoul
Catalog # CMPL498C.1K.W.10
2 COMPOSES A 1 G/L CHAQUE DANS H2O ( Ampoules de 10 ml ) of Formaldehyde [CAS:50-00-0] Acetaldehyde [CAS:75-07-0] 1 * 1 Ampoul
Catalog # CMPL498C.1K.W.10
Frequently Bought Together