ContactUsLinkComponent
Drum 6,5 L stackable with wide mouth and with closure in metal galvanized iron. Material: High density polyethylene, Color: Blue. No ONU. Ø 210 x H 250 x Mouth 175 mm. 1 * 138 items
Catalog # LAMA1368
Supplier:  LAMAPLAST
Drum 6,5 L stackable with wide mouth and with closure in metal galvanized iron. Material: High density polyethylene, Color: Blue. No ONU. Ø 210 x H 250 x Mouth 175 mm. 1 * 138 items
Catalog # LAMA1368
Supplier:  LAMAPLAST
Frequently Bought Together