ContactUsLinkComponent
Blade, scalpel, type 13, 2-sided ground-edged, length:42.3mm, width:6.2mm, material, thickness:0.40mm, single bevel edge-ground. 1 * 10 items
Catalog # MARO13.50
Supplier:  MARTOR ARGENTAX
Blade, scalpel, type 13, 2-sided ground-edged, length:42.3mm, width:6.2mm, material, thickness:0.40mm, single bevel edge-ground. 1 * 10 items
Catalog # MARO13.50
Supplier:  MARTOR ARGENTAX
Frequently Bought Together