ContactUsLinkComponent
High-performance grease for all kinds of greasing points, fine PTFE particles lower the friction between the surfaces, a special additive system provides extremely high pressure and temperature resistance, also suitable as anticorrosive, does not con 1 * 100 g
Catalog # BOHLS1872-16
Supplier:  Bohlender
undefined
High-performance grease for all kinds of greasing points, fine PTFE particles lower the friction between the surfaces, a special additive system provides extremely high pressure and temperature resistance, also suitable as anticorrosive, does not con 1 * 100 g
Catalog # BOHLS1872-16
Supplier:  Bohlender
Frequently Bought Together