ContactUsLinkComponent
15COMPOSES A 50 MG/L DANS ACETONITRILE of Naphthalene [CAS:91-20-3] Acenaphthene [CAS:83-32-9] Fluorene [CAS:86-73-7] Phenanthrene [CAS:85-01-8] Anthracene [CAS:120-12-7] Fluoranthene [CAS:206-44-0] Pyrene [CAS:129-00-0] Benzo(a)anthrac 1 * 5 Ampoul
Catalog # CMPL4790.50.N.1
15COMPOSES A 50 MG/L DANS ACETONITRILE of Naphthalene [CAS:91-20-3] Acenaphthene [CAS:83-32-9] Fluorene [CAS:86-73-7] Phenanthrene [CAS:85-01-8] Anthracene [CAS:120-12-7] Fluoranthene [CAS:206-44-0] Pyrene [CAS:129-00-0] Benzo(a)anthrac 1 * 5 Ampoul
Catalog # CMPL4790.50.N.1
Frequently Bought Together