ContactUsLinkComponent
Capillary column OPTIMA 5 AM ID: 0.25 mm, film thickness: 0.50 µm length: 30 m on 5 inch (13 cm) cage pack of 1 1 * 1 items
Catalog # MANA726354.30E
Supplier:  MACHEREY-NAGEL
Capillary column OPTIMA 5 AM ID: 0.25 mm, film thickness: 0.50 µm length: 30 m on 5 inch (13 cm) cage pack of 1 1 * 1 items
Catalog # MANA726354.30E
Supplier:  MACHEREY-NAGEL
Frequently Bought Together