ContactUsLinkComponent
Vakuwash For rinsing and drying glass cuvettes. Cell is held, open end down, against a rubber cushion which closes vacuum system. Includes 500 mL PP flask, Vikem® stopper, tubing, and Vakuwash funnel 1 * 1 items
Catalog # SIALZ177474-1EA
Supplier:  Merck
Vakuwash For rinsing and drying glass cuvettes. Cell is held, open end down, against a rubber cushion which closes vacuum system. Includes 500 mL PP flask, Vikem® stopper, tubing, and Vakuwash funnel 1 * 1 items
Catalog # SIALZ177474-1EA
Supplier:  Merck
Frequently Bought Together