ContactUsLinkComponent
Recombinant protein, mapk15( aah28034, 1 a.a. - 278 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00225689-P01
Supplier:  Abnova
Recombinant protein, mapk15( aah28034, 1 a.a. - 278 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00225689-P01
Supplier:  Abnova