ContactUsLinkComponent
Recombinant protein, traf7( aah24267.1, 1 a.a. - 594 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00084231-P01
Supplier:  Abnova
Recombinant protein, traf7( aah24267.1, 1 a.a. - 594 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00084231-P01
Supplier:  Abnova