ContactUsLinkComponent
Recombinant protein, ube2q2( np_775740.1, 1 a.a. - 375 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00092912-P01
Supplier:  Abnova
Recombinant protein, ube2q2( np_775740.1, 1 a.a. - 375 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00092912-P01
Supplier:  Abnova