ContactUsLinkComponent
Recombinant protein, nhej1( np_079058.1, 1 a.a. - 299 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00079840-P01
Supplier:  Abnova
Recombinant protein, nhej1( np_079058.1, 1 a.a. - 299 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00079840-P01
Supplier:  Abnova