ContactUsLinkComponent
Recombinant protein, rnf150( aai01993.1, 1 a.a. - 314 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00057484-P01
Supplier:  Abnova
Recombinant protein, rnf150( aai01993.1, 1 a.a. - 314 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00057484-P01
Supplier:  Abnova