Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Zadání objednávky
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CDC42EP4 RECOMBINANT PROTEIN (P01) 1 * 10 µG
ABNOH00023580-P01
:
Abnova
CDC42EP4 RECOMBINANT PROTEIN (P01) 1 * 10 µG
ABNOH00023580-P01
:
Abnova
Literature reference - PubMed ID 25022875
sds_ABNOH00023580-P01.pdf
TDS_ABNOH00023580-P01.pdf
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Sérologická pipeta, standardní
: VWR Collection
Rukavice, nitrilové, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
Acetonitril ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR®, Ubrousky čisticí
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Aceton ≥99%, TECHNICAL
: VWR Chemicals
Sealing film for manual application, Parafilm® M
: HEATHROW
Acetonitril, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® USP, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR®, Pipety Pasteurovy, High performance
: VWR Collection
VWR® NITRILE LIGHT, Rukavice, nitrilové
: VWR Collection