ContactUsLinkComponent
Recombinant protein, mtmr3( np_066576.1, 579 a.a. - 674 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00008897-Q01
Supplier:  Abnova
undefined
Recombinant protein, mtmr3( np_066576.1, 579 a.a. - 674 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00008897-Q01
Supplier:  Abnova