ContactUsLinkComponent
Recombinant protein, pias3( np_006090, 453 a.a. - 550 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00010401-Q01
Supplier:  Abnova
undefined
Recombinant protein, pias3( np_006090, 453 a.a. - 550 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00010401-Q01
Supplier:  Abnova