ContactUsLinkComponent
Recombinant protein, ramp1( np_005846, 27 a.a. - 117 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00010267-Q01
Supplier:  Abnova
Recombinant protein, ramp1( np_005846, 27 a.a. - 117 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00010267-Q01
Supplier:  Abnova