ContactUsLinkComponent
Recombinant protein, trib1( np_079471, 91 a.a. - 191 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00010221-Q01
Supplier:  Abnova
Recombinant protein, trib1( np_079471, 91 a.a. - 191 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00010221-Q01
Supplier:  Abnova