ContactUsLinkComponent
Recombinant protein, pqbp1( np_001027553.1, 91 a.a. - 200 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00010084-Q02
Supplier:  Abnova
Recombinant protein, pqbp1( np_001027553.1, 91 a.a. - 200 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00010084-Q02
Supplier:  Abnova