ContactUsLinkComponent
Recombinant protein, ovgp1( np_002548.3, 579 a.a. - 677 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00005016-Q01
Supplier:  Abnova
Recombinant protein, ovgp1( np_002548.3, 579 a.a. - 677 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00005016-Q01
Supplier:  Abnova