ContactUsLinkComponent
Recombinant protein, sox9( np_000337, 400 a.a. - 509 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00006662-Q01
Supplier:  Abnova
Recombinant protein, sox9( np_000337, 400 a.a. - 509 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00006662-Q01
Supplier:  Abnova