ContactUsLinkComponent
Recombinant protein, nrap( np_932326.2, 1640 a.a. - 1727 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00004892-Q01
Supplier:  Abnova
Recombinant protein, nrap( np_932326.2, 1640 a.a. - 1727 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00004892-Q01
Supplier:  Abnova