ContactUsLinkComponent
Recombinant protein, st6gal1( np_775323, 96 a.a. - 195 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00006480-Q01
Supplier:  Abnova
Recombinant protein, st6gal1( np_775323, 96 a.a. - 195 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00006480-Q01
Supplier:  Abnova