ContactUsLinkComponent
Recombinant protein, rpl3l( np_005052, 280 a.a. - 360 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00006123-Q01
Supplier:  Abnova
undefined
Recombinant protein, rpl3l( np_005052, 280 a.a. - 360 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00006123-Q01
Supplier:  Abnova