ContactUsLinkComponent
Recombinant protein, dao( np_001908, 119 a.a. - 218 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00001610-Q01
Supplier:  Abnova
undefined
Recombinant protein, dao( np_001908, 119 a.a. - 218 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00001610-Q01
Supplier:  Abnova