ContactUsLinkComponent
Recombinant protein, elf2( np_973728, 2 a.a. - 95 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00001998-Q01
Supplier:  Abnova
undefined
Recombinant protein, elf2( np_973728, 2 a.a. - 95 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00001998-Q01
Supplier:  Abnova