ContactUsLinkComponent
Recombinant protein, cd9( aah11988, 112 a.a. - 195 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000928-Q01
Supplier:  Abnova
undefined
Recombinant protein, cd9( aah11988, 112 a.a. - 195 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000928-Q01
Supplier:  Abnova