ContactUsLinkComponent
Recombinant protein, il10( np_000563, 74 a.a. - 178 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00003586-Q01
Supplier:  Abnova
Recombinant protein, il10( np_000563, 74 a.a. - 178 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00003586-Q01
Supplier:  Abnova