ContactUsLinkComponent
Recombinant protein, bmp5( np_066551.1, 323 a.a. - 454 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000653-Q03
Supplier:  Abnova
undefined
Recombinant protein, bmp5( np_066551.1, 323 a.a. - 454 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000653-Q03
Supplier:  Abnova