ContactUsLinkComponent
Recombinant protein, nog( aah34027, 28 a.a. - 233 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00009241-P01
Supplier:  Abnova
Recombinant protein, nog( aah34027, 28 a.a. - 233 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00009241-P01
Supplier:  Abnova