ContactUsLinkComponent
Recombinant protein, gprc5a( np_003970.1, 1 a.a. - 357 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00009052-P01
Supplier:  Abnova
Recombinant protein, gprc5a( np_003970.1, 1 a.a. - 357 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00009052-P01
Supplier:  Abnova